Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr63
Entrez Gene ID
242253
Gene Name
WD repeat domain 63
Gene Aliases
4931433A13Rik
Chromosome Location
Chr.3: 146040526 - 146108107 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172864.3 NP_766452.2 3 - 4 246 132
GenBank mRNA AK029903.1 - 3 - 4 247 132
BC158119.1 - 3 - 4 352 132

View Full Product Details