The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291166.1 | NP_001278095.1 | 4 - 5 | 1284 | 65 |
NM_001291167.1 | NP_001278096.1 | 3 - 4 | 1230 | 65 | |
NM_007461.2 | NP_031487.1 | 4 - 5 | 1281 | 65 | |
GenBank mRNA | BC057620.1 | - | 4 - 5 | 1281 | 65 |
BC060269.1 | - | 1 - 2 | 984 | 65 |