Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Apba2
Entrez Gene ID
11784
Gene Name
amyloid beta (A4) precursor protein-binding, family A, member 2
Gene Aliases
X11-like, X11L, XllL, mXllL
Chromosome Location
Chr.7: 64501706 - 64753878 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291166.1 NP_001278095.1 7 - 8 1549 119
NM_001291167.1 NP_001278096.1 6 - 7 1495 119
NM_007461.2 NP_031487.1 7 - 8 1546 119
GenBank mRNA BC057620.1 - 7 - 8 1546 119
BC060269.1 - 4 - 5 1249 119
L34676.1 - 4 - 5 119

View Full Product Details