Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mtmr10
Entrez Gene ID
233315
Gene Name
myotubularin related protein 10
Gene Aliases
BB128963, C79734
Chromosome Location
Chr.7: 64287670 - 64340806 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172742.2 NP_766330.2 4 - 5 442 85
GenBank mRNA AK088715.1 - 4 - 5 460 85
BC041672.1 - 4 - 5 414 85
BC055074.1 - 2 - 3 106 85
CK634659.1 - 4 - 5 328 85
CN669724.1 - 4 - 5 442 85

View Full Product Details