Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zc3h13
Entrez Gene ID
67302
Gene Name
zinc finger CCCH type containing 13
Gene Aliases
2600010B19Rik, 3110050K21Rik, 4930570G11Rik, C87618
Chromosome Location
Chr.14: 75284308 - 75344433 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026083.2 NP_080359.2 6 - 7 889 132
GenBank mRNA AK014200.1 - 6 - 7 669 132
AK078225.1 - 6 - 7 880 132
BC138264.1 - 6 - 7 838 132

View Full Product Details