The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005846.2 | NP_001005846.1 | 4 - 5 | 632 | 66 |
NM_026656.4 | NP_080932.2 | 4 - 5 | 660 | 66 | |
GenBank mRNA | AF503575.1 | - | 4 - 5 | 649 | 66 |
AK014467.1 | - | 4 - 5 | 563 | 66 | |
AK019454.1 | - | 4 - 5 | 529 | 66 | |
AK150446.1 | - | 4 - 5 | 628 | 66 | |
AK150547.1 | - | 4 - 5 | 628 | 66 | |
AK152067.1 | - | 4 - 5 | 628 | 66 | |
AK152943.1 | - | 4 - 5 | 628 | 66 | |
AK153290.1 | - | 4 - 5 | 628 | 66 | |
AK159172.1 | - | 4 - 5 | 660 | 66 | |
AY083532.1 | - | 3 - 4 | 478 | 66 | |
BC029847.1 | - | 4 - 5 | 509 | 66 |