Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rpgrip1
- Entrez Gene ID
-
77945
- Gene Name
- retinitis pigmentosa GTPase regulator interacting protein 1
- Gene Aliases
-
0610005A07Rik, 4930401L23Rik, 4930505G06Rik, A930002K18Rik, AA415034, nmf247
- Chromosome Location
-
Chr.14: 52104106 - 52161339 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BC055283.1 |
- |
|
6327 |
|
Chr. - - [-] on Build |
133 |
View Full Product Details