The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172601.3 | NP_766189.1 | 7 - 8 | 700 | 145 |
GenBank mRNA | AB232586.1 | - | 7 - 8 | 546 | 145 |
AK005230.1 | - | 7 - 8 | 603 | 145 | |
AK038393.1 | - | 7 - 8 | 635 | 145 | |
AK044202.1 | - | 7 - 8 | 577 | 145 | |
AK160249.1 | - | 7 - 8 | 851 | 145 | |
AK160351.1 | - | 7 - 8 | 566 | 145 | |
BC033312.1 | - | 7 - 8 | 597 | 145 | |
BC046334.1 | - | 7 - 8 | 642 | 145 | |
BC054719.1 | - | 7 - 8 | 633 | 145 |