Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tarsl2
Entrez Gene ID
272396
Gene Name
threonyl-tRNA synthetase-like 2
Gene Aliases
A530046H20Rik
Chromosome Location
Chr.7: 65644898 - 65692093 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172310.2 NP_758514.2 13 - 14 1799 78
GenBank mRNA AK040927.1 - 13 - 14 1772 78
AK041280.1 - 13 - 14 1799 78
AK079416.1 - 13 - 14 1781 78
BC039225.1 - 13 - 14 1783 78

View Full Product Details