Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adam7
Entrez Gene ID
11500
Gene Name
a disintegrin and metallopeptidase domain 7
Gene Aliases
9230104J12Rik, EAP1, EAPI
Chromosome Location
Chr.14: 68497336 - 68533689 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007402.2 NP_031428.2 20 - 21 2218 82
GenBank mRNA AF013107.1 - 20 - 21 2201 82
AK136692.1 - 20 - 21 2217 82
AK164965.1 - 6 - 7 738 82

View Full Product Details