Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbck
Entrez Gene ID
271981
Gene Name
TBC1 domain containing kinase
Gene Aliases
1700120J03Rik, 9430001M19, A630047E20Rik, C030007I09Rik, Tbckl
Chromosome Location
Chr.3: 132683994 - 132841688 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK034533.1 - 20 - 21 2099 96
CN720994.1 - 2 - 3 161 96

View Full Product Details