Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rcbtb1
Entrez Gene ID
71330
Gene Name
regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 1
Gene Aliases
5430409I18Rik, AW111883, CLLD7, CLLL7
Chromosome Location
Chr.14: 59201027 - 59237267 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027764.2 NP_082040.2 1 - 2 357 104
GenBank mRNA AK017287.1 - 1 - 2 103 104
AK033501.1 - 1 - 2 291 104
AK088299.1 - 1 - 2 346 104
CJ183647.1 - 1 - 2 358 104

View Full Product Details