Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Manba
Entrez Gene ID
110173
Gene Name
mannosidase, beta A, lysosomal
Gene Aliases
2410030O07Rik, B930014J03Rik, Bmn
Chromosome Location
Chr.3: 135485575 - 135571404 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027288.3 NP_081564.3 2 - 3 341 84
GenBank mRNA AF306557.1 - 2 - 3 345 84
AK036505.1 - 2 - 3 339 84
AK047057.1 - 2 - 3 334 84
AK142165.1 - 2 - 3 306 84
AK154172.1 - 2 - 3 319 84
AK166022.1 - 2 - 3 323 84
AK166340.1 - 2 - 3 339 84
AK170290.1 - 2 - 3 338 84
AK171416.1 - 2 - 3 328 84
BC031409.1 - 2 - 3 284 84

View Full Product Details