Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pnma2
Entrez Gene ID
239157
Gene Name
paraneoplastic antigen MA2
Gene Aliases
A830049P17Rik, mKIAA0883
Chromosome Location
Chr.14: 66900682 - 66920063 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175498.4 NP_780707.1 1 - 2 85 106
GenBank mRNA AK043718.1 - 1 - 2 33 106
AK220357.1 - - 40 106

View Full Product Details