Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Xpo4
Entrez Gene ID
57258
Gene Name
exportin 4
Gene Aliases
B430309A01Rik, mKIAA1721
Chromosome Location
Chr.14: 57577520 - 57664985 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020506.1 NP_065252.1 19 - 20 2921 144
GenBank mRNA AF145021.1 - 19 - 20 2921 144
AK042783.1 - 2 - 3 263 144
AK129430.1 - 19 - 20 2916 144

View Full Product Details