Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mark4
Entrez Gene ID
232944
Gene Name
MAP/microtubule affinity regulating kinase 4
Gene Aliases
2410090P21Rik, C79806, Markl1
Chromosome Location
Chr.7: 19425102 - 19458795 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172279.1 NP_758483.1 6 - 7 490 100
GenBank mRNA AK122565.2 - 2 - 3 136 100
AK146784.1 - 7 - 8 825 100
AY151083.1 - 6 - 7 490 100

View Full Product Details