The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010836.3 | NP_001010836.1 | 7 - 8 | 1469 | 97 |
GenBank mRNA | AK146098.1 | - | 7 - 8 | 1445 | 97 |
AK170535.1 | - | 7 - 8 | 1470 | 97 | |
AY869711.1 | - | 7 - 8 | 1439 | 97 |