The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010836.3 | NP_001010836.1 | 12 - 13 | 2563 | 67 |
GenBank mRNA | AK146098.1 | - | 12 - 13 | 2542 | 67 |
AK170535.1 | - | 12 - 13 | 2564 | 67 | |
AY869711.1 | - | 12 - 13 | 2533 | 67 |