The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253812.1 | NP_001240741.1 | 10 - 11 | 1267 | 67 |
NM_028811.3 | NP_083087.1 | 10 - 11 | 1210 | 67 | |
NR_045599.1 | 10 - 11 | 1210 | 67 | ||
GenBank mRNA | AK012072.1 | - | 10 - 11 | 1148 | 67 |
AK088457.1 | - | 10 - 11 | 1231 | 67 | |
AK150058.1 | - | 10 - 11 | 1148 | 67 | |
BC026461.1 | - | 8 - 9 | 862 | 67 | |
BC037470.1 | - | 4 - 5 | 564 | 67 | |
BC048822.1 | - | 5 - 6 | 715 | 67 | |
BC057453.1 | - | 10 - 11 | 1150 | 67 |