Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gulo
Entrez Gene ID
268756
Gene Name
gulonolactone (L-) oxidase
Gene Aliases
5730581M22, AU018375, BC028822, GLO, LGO, sfx, unh, unhip
Chromosome Location
Chr.14: 65986786 - 66009254 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178747.3 NP_848862.1 7 - 8 876 76
GenBank mRNA AK077740.1 - 7 - 8 877 76
AK167460.1 - 7 - 8 865 76
AY453064.1 - 6 - 7 713 76
BC019856.1 - 7 - 8 823 76
BC028822.1 - 7 - 8 832 76
BC028828.1 - 7 - 8 835 76
BC034835.1 - 6 - 7 632 76

View Full Product Details