Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adam2
Entrez Gene ID
11495
Gene Name
a disintegrin and metallopeptidase domain 2
Gene Aliases
AI323749, Ftnb, Ph30-beta
Chromosome Location
Chr.14: 66027329 - 66077856 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009618.2 NP_033748.2 5 - 6 368 142
GenBank mRNA AK016550.1 - 5 - 6 368 142
AK161261.1 - 5 - 6 333 142
AK161266.1 - 5 - 6 357 142
U16242.1 - 5 - 6 357 142
U22057.1 - 4 - 5 308 142

View Full Product Details