The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029979.3 | NP_084255.2 | 5 - 6 | 990 | 67 |
GenBank mRNA | AB060155.1 | - | 5 - 6 | 934 | 67 |
AF145374.1 | - | 5 - 6 | 974 | 67 | |
AK003000.1 | - | 5 - 6 | 949 | 67 | |
AK032635.1 | - | 5 - 6 | 963 | 67 | |
AK075877.1 | - | 5 - 6 | 961 | 67 | |
AK146541.1 | - | 4 - 5 | 441 | 67 | |
BC049105.1 | - | 5 - 6 | 940 | 67 | |
BC053494.1 | - | 5 - 6 | 855 | 67 | |
BC145783.1 | - | 5 - 6 | 984 | 67 | |
BC145809.1 | - | 5 - 6 | 984 | 67 |