The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164171.1 | NP_001157643.1 | 3 - 4 | 404 | 61 |
NM_010856.4 | NP_034986.1 | 3 - 4 | 340 | 61 | |
GenBank mRNA | AK142232.1 | - | 3 - 4 | 377 | 61 |
BC061145.1 | - | 3 - 4 | 373 | 61 | |
BC090979.1 | - | 3 - 4 | 373 | 61 | |
BC110700.1 | - | 3 - 4 | 386 | 61 | |
BC138027.1 | - | 2 - 3 | 279 | 61 | |
DQ023232.1 | - | 3 - 4 | 373 | 61 | |
DQ023233.1 | - | 3 - 4 | 438 | 61 | |
DQ023234.1 | - | 3 - 4 | 404 | 61 | |
DQ023235.1 | - | 3 - 4 | 305 | 61 | |
DQ023236.1 | - | 3 - 4 | 338 | 61 | |
DQ023237.1 | - | 3 - 4 | 328 | 61 | |
M76598.1 | - | 1 - 2 | 202 | 61 | |
M76599.1 | - | 1 - 2 | 202 | 61 | |
M76600.1 | - | 1 - 2 | 202 | 61 | |
M76601.1 | - | 3 - 4 | 304 | 61 |