The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164503.1 | NP_001157975.1 | 4 - 5 | 424 | 95 |
GenBank mRNA | AK158300.1 | - | 4 - 5 | 345 | 95 |