The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144817.2 | NP_659066.1 | 12 - 13 | 1595 | 140 |
GenBank mRNA | AF428262.1 | - | 12 - 13 | 1543 | 140 |
AY212936.1 | - | 11 - 12 | 1475 | 140 | |
BC021840.1 | - | 12 - 13 | 1544 | 140 |