Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Exoc5
Entrez Gene ID
105504
Gene Name
exocyst complex component 5
Gene Aliases
AI447711, AI448003, Gm76, PRO1912, SEC10, Sec10l1
Chromosome Location
Chr.14: 49011922 - 49066667 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207214.3 NP_997097.1 12 - 13 1806 149
GenBank mRNA AK134562.1 - 12 - 13 1319 149
AK150297.1 - 1 - 2 73 149
AK152307.1 - 12 - 13 1535 149
AK157473.1 - 12 - 13 1501 149
AK164067.1 - 12 - 13 1528 149
AK167145.1 - 12 - 13 1494 149
AK170148.1 - 12 - 13 1500 149
AK171012.1 - 12 - 13 1524 149
BC049967.1 - 12 - 13 1463 149

View Full Product Details