Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serp2
Entrez Gene ID
72661
Gene Name
stress-associated endoplasmic reticulum protein family member 2
Gene Aliases
2810032E02Rik
Chromosome Location
Chr.14: 76532812 - 76557919 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027699.1 4 - 5 468 65
GenBank mRNA BC100483.1 - 4 - 5 437 65

View Full Product Details