Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atxn7
Entrez Gene ID
246103
Gene Name
ataxin 7
Gene Aliases
A430107N12Rik, AI627028, Sca7, ataxin-7
Chromosome Location
Chr.14: 13961439 - 14107302 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF455111.1 - 4 - 5 726 75
BC137846.1 - 5 - 6 907 75

View Full Product Details