Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psmd6
Entrez Gene ID
66413
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 6
Gene Aliases
2400006A19Rik
Chromosome Location
Chr.14: 14112185 - 14120904 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025550.2 NP_079826.2 1 - 2 170 76
GenBank mRNA AJ252147.1 - 1 - 2 206 76
AK002343.1 - 1 - 2 204 76
AK010287.1 - 1 - 2 203 76
BC006869.1 - 1 - 2 170 76

View Full Product Details