The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153136.4 | NP_694776.2 | 1 - 2 | 260 | 58 |
GenBank mRNA | AK137014.1 | - | 1 - 2 | 471 | 58 |
AK145997.1 | - | 1 - 2 | 249 | 58 | |
AK155086.1 | - | 1 - 2 | 261 | 58 | |
AK157937.1 | - | 1 - 2 | 261 | 58 |