The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010910.1 | NP_035040.1 | 1 - 2 | 1129 | 69 |
GenBank mRNA | AK149219.1 | - | - | 1160 | 69 |
BC016436.1 | - | 1 - 2 | 1130 | 69 | |
BC029203.1 | - | 1 - 2 | 1129 | 69 | |
DQ201635.1 | - | 1 - 2 | 1106 | 69 | |
M20480.1 | - | 1 - 2 | 1076 | 69 | |
X02165.1 | - | 1 - 2 | 329 | 69 |