Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- 4933429O19Rik
- Entrez Gene ID
-
66770
- Gene Name
- RIKEN cDNA 4933429O19 gene
- Gene Aliases
-
1700031B11Rik
- Chromosome Location
-
Chr.14: 48878873 - 48887112 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033783.1 |
|
1 - 2 |
241 |
60 |
GenBank mRNA |
AK006575.1 |
- |
1 - 2 |
243 |
60 |
AK015198.1 |
- |
1 - 2 |
197 |
60 |
AK016985.1 |
- |
1 - 2 |
231 |
60 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BY716669.1 |
- |
1 - 2 |
231 |
|
Chr. 14 - 48878884 - 48886191 [+] on Build GRCm38 |
60 |
GenBankmRNA |
BU937300.1 |
- |
|
212 |
|
Chr. - - [-] on Build |
60 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
orthodenticle homolog 2 opposite strand 1 |
Otx2os1 |
|
-85.5kb
|
Sense |
Chr. - 48669288 - 48793348 on Build GRCm38 |
Downstream |
predicted gene, 36024 |
Gm36024 |
|
101.6kb
|
Anti-sense |
Chr. - 48982480 - 48984380 on Build GRCm38 |
View Full Product Details