The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113532.1 | NP_001107004.1 | 1 - 2 | 187 | 102 |
NM_001113533.1 | NP_001107005.1 | 1 - 2 | 187 | 102 | |
GenBank mRNA | AK156334.1 | - | 1 - 2 | 83 | 102 |
BC046416.1 | - | 1 - 2 | 116 | 102 | |
BC047338.1 | - | 1 - 2 | 116 | 102 | |
BC093504.1 | - | 1 - 2 | 120 | 102 | |
BY016042.1 | - | 1 - 2 | 187 | 102 |