The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197046.1 | NP_001183975.1 | 3 - 4 | 291 | 91 |
NM_201230.5 | NP_957682.1 | 3 - 4 | 291 | 91 | |
GenBank mRNA | AK016110.1 | - | 3 - 4 | 294 | 91 |
AK028778.1 | - | 3 - 4 | 220 | 91 | |
AK029470.1 | - | 3 - 4 | 290 | 91 | |
AK031261.1 | - | 3 - 4 | 274 | 91 | |
AK050389.1 | - | 3 - 4 | 229 | 91 | |
BC050774.1 | - | 3 - 4 | 243 | 91 | |
BC058804.1 | - | 3 - 4 | 226 | 91 | |
BC080717.1 | - | 3 - 4 | 278 | 91 | |
BC108331.1 | - | 3 - 4 | 289 | 91 |