Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arhgef40
Entrez Gene ID
268739
Gene Name
Rho guanine nucleotide exchange factor (GEF) 40
Gene Aliases
E130112L23Rik, Gm669, Solo
Chromosome Location
Chr.14: 51984681 - 52006247 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145921.1 NP_001139393.1 7 - 8 2068 70
NM_001145922.1 NP_001139394.1 7 - 8 2068 70
NM_198249.4 NP_937892.2 7 - 8 2068 70
GenBank mRNA AK131136.1 - 7 - 8 2037 70
AK143527.1 - 7 - 8 2043 70
AK147761.1 - 7 - 8 2004 70
AK163945.1 - 7 - 8 2069 70
BC059018.1 - 7 - 8 1970 70

View Full Product Details