Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fhit
Entrez Gene ID
14198
Gene Name
fragile histidine triad gene
Gene Aliases
AW045638, Fra14A2
Chromosome Location
Chr.14: 9550092 - 11162054 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AA797426.1 - 2 - 3 126 62
AF047699.1 - 5 - 6 621 62
AF055573.1 - - 487 62
BC012662.1 - 3 - 4 346 62
CO040033.1 - 3 - 4 62

View Full Product Details