Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grm3
Entrez Gene ID
108069
Gene Name
glutamate receptor, metabotropic 3
Gene Aliases
0710001G23Rik, Gprc1c, mGlu3, mGluR3
Chromosome Location
Chr.5: 9485236 - 9725352 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181850.2 NP_862898.1 5 - 6 2937 78
GenBank mRNA AK002958.1 - 2 - 3 1141 78
BC060060.1 - 3 - 4 1295 78
BC111891.1 - 5 - 6 2755 78
BC113170.1 - 5 - 6 2755 78

View Full Product Details