Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ephx1
Entrez Gene ID
13849
Gene Name
epoxide hydrolase 1, microsomal
Gene Aliases
AI195553, Eph-1, Eph1, Epxh1, mEH
Chromosome Location
Chr.1: 180989556 - 181017495 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010145.2 NP_034275.1 1 - 2 75 71
GenBank mRNA AK018249.1 - 1 - 2 84 71
BC029636.1 - 1 - 2 74 71
BC045194.1 - 1 - 2 75 71
BC057857.1 - 1 - 2 70 71
BC061493.1 - 1 - 2 101 71

View Full Product Details