Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lmo4
Entrez Gene ID
16911
Gene Name
LIM domain only 4
Gene Aliases
A730077C12Rik, Crp3, Etohi4
Chromosome Location
Chr.3: 144188530 - 144205255 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161769.1 NP_001155241.1 3 - 4 489 66
NM_001161770.1 NP_001155242.1 2 - 3 621 66
NM_010723.3 NP_034853.1 3 - 4 776 66
GenBank mRNA AF074600.1 - 2 - 3 337 66
AF096996.1 - 3 - 4 505 66
AF102817.1 - 3 - 4 406 66
AK080534.1 - 3 - 4 424 66
AK138278.1 - 2 - 3 622 66
AK159590.1 - 3 - 4 703 66
BC003488.1 - 2 - 3 488 66
BC004661.1 - 3 - 4 429 66
BC010278.1 - 3 - 4 519 66
BC100382.1 - 3 - 4 381 66

View Full Product Details