Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rabggtb
Entrez Gene ID
19352
Gene Name
Rab geranylgeranyl transferase, b subunit
Gene Aliases
-
Chromosome Location
Chr.3: 153907289 - 153912966 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163478.1 NP_001156950.1 8 - 9 884 68
NM_001163479.1 NP_001156951.1 6 - 7 822 68
NM_011231.2 NP_035361.2 7 - 8 1022 68
NR_028094.1 9 - 10 966 68
NR_028095.1 8 - 9 1104 68
NR_028096.1 8 - 9 1120 68
NR_028097.1 7 - 8 904 68
NR_028098.1 6 - 7 806 68
GenBank mRNA AF231926.1 - 8 - 9 960 68
AK010690.1 - 6 - 7 574 68
AK159902.1 - 9 - 10 1330 68
AK160162.1 - 8 - 9 871 68
BC003948.1 - 8 - 9 947 68
BC021335.1 - 7 - 8 868 68
BC057661.1 - 7 - 8 1536 68
BC132473.1 - 7 - 8 900 68
BC138547.1 - 7 - 8 900 68
BC145323.1 - 8 - 9 982 68
BC145324.1 - 7 - 8 884 68
BC145325.1 - 8 - 9 998 68
BC145326.1 - 8 - 9 958 68
U12922.1 - 7 - 8 878 68

View Full Product Details