The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_028094.1 | 7 - 8 | 709 | 102 | |
NR_028095.1 | 6 - 7 | 847 | 102 | ||
NR_028097.1 | 5 - 6 | 647 | 102 | ||
NR_028098.1 | 4 - 5 | 549 | 102 | ||
GenBank mRNA | AF231926.1 | - | 6 - 7 | 703 | 102 |
BC003948.1 | - | 6 - 7 | 690 | 102 | |
BC057661.1 | - | 5 - 6 | 1279 | 102 | |
BC145323.1 | - | 6 - 7 | 725 | 102 | |
BC145324.1 | - | 5 - 6 | 627 | 102 | |
BG076913.2 | - | 4 - 5 | 318 | 102 | |
CV562676.1 | - | 5 - 6 | 493 | 102 |