Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rwdd3
Entrez Gene ID
66568
Gene Name
RWD domain containing 3
Gene Aliases
2510027J23Rik, 3110037C01Rik, AI035684, AI662458, Rsume, X2CR1
Chromosome Location
Chr.3: 121155400 - 121171780 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028456.3 NP_082732.2 1 - 2 105 84
GenBank mRNA AF439556.1 - 1 - 2 104 84
AK014130.1 - 1 - 2 43 84
AK134288.1 - 1 - 2 96 84
BC128287.1 - 1 - 2 104 84
BC139340.1 - 1 - 2 104 84
BC139342.1 - 1 - 2 104 84

View Full Product Details