Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pla2g12a
Entrez Gene ID
66350
Gene Name
phospholipase A2, group XIIA
Gene Aliases
2310004B05Rik, GXII, GXII-1, GXII-1-PLA2, Pla2g12, Rossy, mGXII-1
Chromosome Location
Chr.3: 129878606 - 129895825 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183423.3 NP_904359.1 1 - 2 116 88
GenBank mRNA AY007382.1 - 1 - 2 116 88

View Full Product Details