Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mphosph8
Entrez Gene ID
75339
Gene Name
M-phase phosphoprotein 8
Gene Aliases
1500035L22Rik, 4930548G07Rik, AU040635
Chromosome Location
Chr.14: 56668240 - 56697430 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023773.2 NP_076262.2 3 - 4 1317 153
GenBank mRNA AB041598.2 - 3 - 4 1318 153
AK158773.1 - 3 - 4 1295 153
AK159495.1 - 3 - 4 1295 153
BC145164.1 - 3 - 4 1242 153
BC145700.1 - 3 - 4 1242 153

View Full Product Details