Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pigk
Entrez Gene ID
329777
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class K
Gene Aliases
3000001O05Rik
Chromosome Location
Chr.3: 152714100 - 152789015 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025662.5 NP_079938.1 2 - 3 184 93
NM_178016.3 NP_821135.1 2 - 3 184 93
GenBank mRNA AK013853.1 - 2 - 3 181 93
AK043033.1 - - 173 93
AK046246.1 - 2 - 3 172 93
AK077893.1 - 2 - 3 185 93
AK082806.1 - 2 - 3 177 93
AK160801.1 - 2 - 3 179 93
AK168666.1 - - 166 93
BC023256.1 - 2 - 3 166 93
BC060175.1 - 2 - 3 170 93

View Full Product Details