Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc44a5
Entrez Gene ID
242259
Gene Name
solute carrier family 44, member 5
Gene Aliases
AA117069, Gm422
Chromosome Location
Chr.3: 153973436 - 154271747 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081263.1 NP_001074732.1 18 - 19 2112 105
GenBank mRNA BC068154.1 - 5 - 6 897 105
BC086641.1 - 18 - 19 2112 105

View Full Product Details