Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cpne6
Entrez Gene ID
12891
Gene Name
copine VI
Gene Aliases
AU067659, BB076446
Chromosome Location
Chr.14: 55510063 - 55517431 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136057.2 NP_001129529.1 3 - 4 637 68
NM_001146183.1 NP_001139655.1 3 - 4 614 68
NM_009947.3 NP_034077.1 3 - 4 440 68
GenBank mRNA AB008893.1 - 3 - 4 440 68
AK134548.1 - 3 - 4 459 68
AK144410.1 - 3 - 4 466 68
AK158638.1 - 2 - 3 1859 68
BC050766.1 - 3 - 4 462 68
BC051994.1 - 2 - 3 1022 68

View Full Product Details