Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
R3hcc1
Entrez Gene ID
71843
Gene Name
R3H domain and coiled-coil containing 1
Gene Aliases
1700020M16Rik, AA407796
Chromosome Location
Chr.14: 69697304 - 69714809 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146012.2 NP_001139484.2 4 - 5 1092 94
NM_001301651.1 NP_001288580.1 4 - 5 1044 94
NM_001301652.1 NP_001288581.1 3 - 4 1325 94
GenBank mRNA AK006177.1 - 4 - 5 1044 94
AK032854.1 - 4 - 5 1062 94
AK156843.1 - 4 - 5 1062 94
BC052536.1 - 3 - 4 1241 94
BC059860.1 - 3 - 4 1325 94

View Full Product Details