Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnaseh2b
Entrez Gene ID
67153
Gene Name
ribonuclease H2, subunit B
Gene Aliases
1110019N06Rik, 2610207P08Rik, Dleu8
Chromosome Location
Chr.14: 62332068 - 62392708 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026001.2 NP_080277.1 7 - 8 618 69
GenBank mRNA AK011914.1 - 7 - 8 657 69
AK082752.1 - 7 - 8 619 69
BC047997.1 - 7 - 8 619 69

View Full Product Details