The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026001.2 | NP_080277.1 | 10 - 11 | 820 | 91 |
GenBank mRNA | AK011914.1 | - | 10 - 11 | 859 | 91 |
AK082752.1 | - | 10 - 11 | 821 | 91 | |
BC047997.1 | - | 10 - 11 | 821 | 91 |